missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AmnioMAX™ C-100 Complete Medium

Product Code. 10620186 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10620186 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10620186 Supplier Gibco™ Supplier No. 12558011
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Part of kit used to prepare fully supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures

Gibco™ AmnioMAX™ C-100 Complete Medium is a kit used to prepare a fully-supplemented medium developed for the short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures.

Gibco™ AmnioMAX™ C-100 Complete Medium features:

  • Easy-to-use format
  • Quality and performance testing
  • Unique, optimized formulation

Easy-to-use format

Gibco™ AmnioMAX™ C-100 Complete Medium is a two-part kit for preparation of a complete medium that requires no additional supplementation. The complete medium can be stored at 2–8°C for up to ten days.

Quality and performance testing

Every lot of Gibco™ AmnioMAX™ C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Pooled primary human amniotic fluid samples are cultured for six days in Gibco™ AmnioMAX™ C-100 Complete Medium before measuring the total number of colonies and total number of mitotic colonies. In addition, each lot is tested for pH and osmolality and must pass a sterility test prior to lot release.

Unique, optimized formulation

Gibco™ AmnioMAX™ C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth. This optimized medium also has an enhanced buffering system that provides greater pH stability during culture manipulations.

Product Use

For in vitro diagnostic use.

cGMP Manufacturing and Quality System

Gibco™ AmnioMAX™ C-100 Complete Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Compliance

Gibco AmnioMAX™ C-100 Complete Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Amniocyte Cells
Form Liquid
Product Type Complete Medium
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives Glutamine
Product Line AmnioMAX
Quantity 1 Set
Green Features ACT Ecolabel (Silver)
Included Antibiotics Gentamicin
Content And Storage Basal Medium (90 ml):
Storage: 2 – 8°C
Ships: Ambient temperature
Shelf life: 16 months from date of manufacture

Supplement (15 ml):
Storage: -5 to -20°C
Ships: Frozen
Shelf life: 16 months from date of manufacture

Use complete medium within 7–10 days once opened.
Show More Show Less

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.