missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Carbenicillin Disodium Salt

Product Code. 11568616
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11568616

Brand: Gibco™ 10177012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semi-synthetic penicillin antibiotic

Carbenicillin Disodium Salt is a semi-synthetic penicillin antibiotic which interferes with cell wall synthesis of gram-negative bacteria while displaying low toxicity to plant tissues.

  • Used as selective antibiotic for resistant Agrobacterium and E. coli, generally at a concentration of 100 - 500μg/mL
  • Provided as powder and should be made into stock solution at 100 – 500mg/mL in water

Cloning, Transformation

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Color White, off-white
Content And Storage Storage conditions: 2 to 8°C
Shipping conditions: Ice
Shelf life: 36 months from date of manufacture
Concentration 50 to 100 μg/mL
Form Powder
Product Type Antibiotic
Sterility Sterile
Product Line Gibco
Quantity 5 g
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.