missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, powder, calcium, magnesium

Product Code. p-4924955
Click to view available options
Quantity:
1 x 10 L
Unit Size:
10 L
This item is not returnable. View return policy

Product Code. 11510566

Brand: Gibco™ 21300058

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:
With: Calcium, Magnesium
Without: Phenol Red

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
For Use With (Application) Mammalian Cell Culture
Physical Form Powder
Product Line Gibco
Quantity 1 x 10 L
Solution Type Dulbecco's Phosphate Buffered Saline
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard

For in vitro diagnostic use. Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.