missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, powder, no calcium, no magnesium

Product Code. 11560576
Change view
Click to view available options
Quantity:
10 L
10 x 1 L
Unit Size:
10 L
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11560576 10 L 10 L
10443832 10 x 1 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11560576 Supplier Gibco™ Supplier No. 21600069

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:

  • Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
For Use With (Application) Mammalian Cell Culture
pH 7.2 to 7.7
Physical Form Powder
Product Line Gibco
Quantity 10 L
Solution Type Dulbecco's Phosphate Buffered Saline
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 265 - 310 mOsm/kg

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.