missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ EBSS, no calcium, no magnesium, no phenol red
GREENER_CHOICE

Product Code. 12539069
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12539069

Brand: Gibco™ 14155063

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Gibco™ EBSS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.

This EBSS is manufactured as follows:

With: Glucose, Sodium Bicarbonate

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Earle's Balanced Salt
Concentration 1 X
For Use With (Application) Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue Samples; Diluting Cells for Counting; and Preparing Reagents
pH 7.0 - 8.1
Physical Form Liquid
Product Line Gibco™
Quantity 500 mL
Solution Type Earle's Balanced Salt
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 270 - 310 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.