missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FreeStyle™ 293 Expression Medium

Product Code. 13426156
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
Unit Size:
10 L
1000mL
20 L
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13426156 10 L 10 L
11550426 1 L 1000mL
10319322 6 x 1 L Pack of 6
15285885 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13426156 Supplier Gibco™ Supplier No. 12338001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An animal origin-free, serum-free, protein-free formulation, allowing for easier purification

Gibco FreeStyle 293 Expression Medium is an animal origin–free, chemically defined, protein-free medium specifically developed to support the growth and transfection of 293-F cells in suspension without adaptation. The medium is complete and ready-to-use, with no supplementation required.

Features of FreeStyle 293 Expression Medium include:

  • Complete and ready-to-use format containing GlutaMAX supplement
  • Ability to support suspension growth and transfection of 293-F cells
  • Animal origin–free, protein-free and chemically defined formulation
  • Easy use with the FreeStyle 293 expression systems
  • Scalability in spinner flasks and bioreactors

Complete and ready-to-use format containing GlutaMAX supplement

FreeStyle 293 Expression Medium is ready to use. It does not require the addition of serum, glutamine, or surfactants. It contains GlutaMAX supplement to minimize toxic ammonia build-up and improve cell viability and growth in an easy-to-use format.

Ability to support suspension growth and transfection of 293-F cells

Gibco 293-F, 293-H, and FreeStyle 293-F cells in suspension can be cultured directly into FreeStyle 293 Expression Medium without adaptation. Cells adapted to other commercially available serum-free media can be subcultured directly into FreeStyle 293 Expression Medium, usually without any further adaptation. Cells usually require adaptation from serum-containing formulations. Cells cultured in FreeStyle 293 Expression Medium can be transfected using lipid based transfection reagents, such as 293fectin reagent (Cat. No. 12347019) and Freestyle MAX reagent (16447500).

Animal origin–free, protein-free and chemically defined formulation

FreeStyle 293 Expression Medium is animal origin–free, protein-free and chemically defined, allowing for easier purification of your protein of interest. Gibco chemically defined media contain no proteins, hydrolysates, or components of unknown composition.

Easy use as part of the FreeStyle 293 Expression System and FreeStyle MAX Expression System for protein production

The FreeStyle 293 Expression System (Cat. No. K900001) is a complete, suspension cell culture system for generating large amounts of mammalian recombinant protein (see figure). It combines FreeStyle 293 Expression Medium with 293fectin reagent, a cationic-lipid formulation designed to transfect suspension FreeStyle 293-F cells at high efficiency. The FreeStyle MAX Expression System is a breakthrough technology for rapid and high-yield mammalian protein production. It combines FreeStyle media, FreeStyle MAX Reagent, and either FreeStyle CHO-S cells or FreeStyle 293-F cells.

Scalability in spinner flasks and bioreactors

Protein production using FreeStyle 293 Expression Medium can be scaled up in spinner flasks or bioreactors. The appropriate spinner or impeller speed and seeding density should be optimized for each system. Depending on the impeller design and speed, it may be necessary to supplement the medium with additional Pluronic F-68 to avoid sheer stress in the culture.

Universal bag packaging

Gibco universal bags offer flexible packaging that allows you to integrate a single-use, disposable large volume media container with a wide variety of automated cell culture instruments, disposable bioreactor systems, and sterile connection processes, minimizing the need for costly customization.

Product use

Customers using FreeStyle 293 Expression Medium in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system

FreeStyle 293 Expression Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line FreeStyle™ 293-F Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1X
Form Liquid
Product Type Expression Medium
Serum Level Serum-free
Sterility Sterile
With Additives Non Essential Amino Acids (NEAA), Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Phenol Red
For Use With (Application) Antibody and cell culture production, mammalian cell culture, protein expression, viral vaccine production
Product Line Freestyle
Quantity 10 L
Shipping Condition Room Temperature
Certifications/Compliance ISO 13485 standards, FDA
Endotoxin Level Low
Format Universal Bag
Included Antibiotics No antibiotics
pH 7 to 7.6
Purity or Quality Grade Cell Culture Grade
Regulatory Status For research use only
Concentration Ratio 1:1
Content And Storage Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Culture Environment CO2
Description FreeStyle™ 293 Expression Medium, 10L, Liquid Form, With Sodium Pyruvate, 7 to 7.6 pH, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Soluble in Water, Used for Antibody and Cell Culture Production, Mammalian Cell Culture
Culture Type Suspension Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.