Learn More
Gibco™ Blasticidin S HCl (10 mg/mL)
Description
Blasticidin S is a nucleoside antibiotic isolated from Streptomyces griseochromogenes. It is a potent inhibitor of protein synthesis in both prokaryotic and eukaryotic cells. Blasticidin S acts quickly by inhibiting peptide bond formation by the ribosome.
- Used for prevention of contamination of cell cultures
- Recommended working concentration ranges 1–20μg/mL depending on cell line
- Provided as solution at 10mg/mL in 20 mM HEPES (pH 7.2–7.5)
- Supplied in 10 vials each containing 1mL.
- Resistance is conferred by expression of either one of two Blasticidin S deaminase genes: BSD or bsr
Cell Culture, Cloning, Drosophila S2 Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 and Sf21 Cell Culture, Transfection, Transformation
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | Storage conditions: -5°C to -20°C (protect from light) Shipping conditions: Dry ice Shelf life: 9 months from date of manufacture |
| Cell Type | Eukaryotic Cells, Prokaryotic Cells |
| Concentration | 10 mg/mL |
| Culture Type | Mammalian Cell Culture, Insect Cell Culture |
| Form | Liquid |
| Product Type | Antibiotic |
| Sterility | Sterile-filtered |
| With Additives | HEPES |
| Product Line | Gibco |
| Quantity | 20 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.