missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Glasgow's MEM (GMEM)

Product Code. 15400564 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400564 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15400564 Supplier Gibco™ Supplier No. 11710035
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Supplementation

Glasgow's MEM (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's Minimal Essential Medium.

Glasgow's MEM (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's Minimal Essential Medium. It was used to study the genetic factors that affected cell competence. Glasgow's MEM was developed for use with kidney cell lines, such as BHK-21.

This Glasgow's MEM is modified as follows:

With: L-glutamine, Phenol Red

Without: HEPES, Tryptose Phosphate Broth

The complete formulation is available.

Using Glasgow's MEM

Glasgow's MEM is unique from other media as it contains twice the concentration of amino acids and vitamins compared to the original Basal Medium Eagle, and is used without serum. Glasgow's MEM was originally formulated with 10% tryptose phosphate broth. Glasgow's MEM contains no proteins, lipids, or growth factors. Therefore, Glasgow's MEM requires supplementation with 10% tryptose phosphate broth. Glasgow's MEM uses a sodium bicarbonate buffer system (2.75 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line BHK-21
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type MEM (Minimum Essential Medium)
Sterility Sterile-filtered
With Additives High Glucose, Glutamine, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate, No Tryptose Phosphate Broth
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.