missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, no calcium, no magnesium
GREENER_CHOICE

Product Code. 12082739
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12082739 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12082739 Supplier Gibco™ Supplier No. 14170112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Phenol Red, Glucose

Without: Calcium, Magnesium

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (HBSS)
Concentration 1 X
For Use With (Application) Mammalian Cell Culture
pH 6.7 to 7.8
Physical Form Liquid
Product Line Gibco
Quantity 500 mL
Solution Type Hank's Balanced Salt
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 235 - 295 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.