missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A (Modified) Medium, HEPES

Product Code. 15470574 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470574 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470574 Supplier Gibco™ Supplier No. 12330031
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Supplementation

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues.

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

This McCoy's 5A is modified as follows:

With: High Glucose, L-glutamine, Bacto-peptone, Phenol Red, HEPES

Without: Sodium Pyruvate

The complete formulation is available.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Rat fibroblasts
Cell Type Biopsy Tissues
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type McCoy's 5A Modified Medium
Sterility Sterile-filtered
With Additives High Glucose, Glutamine, HEPES, Phenol Red
Without Additives No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For human ex vivo tissue and cell culture processing applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.