Learn More
Gibco™ MEM, NEAA, powder
Description
Requires
Supplementation
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This MEM is modified as follows:
With: NEAA, L-glutamine, Phenol Red
Without: HEPES, Sodium Bicarbonate
The complete formulation is available.
Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle's salts.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Form | Powder |
| Serum Level | Standard Serum Supplementation |
| With Additives | Low Glucose, Glutamine, Phenol Red |
| Without Additives | No HEPES, No Sodium Pyruvate |
| Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
| Product Line | Gibco |
| Quantity | 50 L |
| Shipping Condition | Room Temperature |
| Content And Storage | Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.