missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM α, nucleosides, no phenol red
GREENER_CHOICE

Product Code. 11534476
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11534476 500 mL 500mL
15673854 10 x 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11534476 Supplier Gibco™ Supplier No. 41061029
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Requires

Supplementation

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells.

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM α is modified as follows:

With: Ribonucleosides, Deoxyribonucleosides, L-glutamine

Without: Phenol Red

The complete formulation is available.

Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. This product is made with Earle's salts.

Product Use

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco MEM α is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type MEM α (Minimum Essential Medium α)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Low Glucose, Glutamine, Sodium Pyruvate, Ribonucleosides, Deoxyribonucleosides
Without Additives No HEPES, No Phenol Red
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.