missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gentamicin (50 mg/mL)

Product Code. 12519079 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 mL
10 x 10 mL
Unit Size:
100mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12519079 10 x 10 mL 100mL
12664735 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12519079 Supplier Gibco™ Supplier No. 15750078
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea

  • Gentamicin sulfate is a water-soluble antibiotic originally purified from the fungus Micromonospora purpurea
  • It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria
  • Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used to prevent the contamination of cell cultures by bacteria
  • The recommended working concentration ranges from 0.5 to 50 μg /mL
  • Store at 15°C to 30°C

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Type Gentamicin
Physical Form Liquid
Sterile Filtered Yes
Concentration 50 mg/mL
Quantity 10 x 10 mL
Validated Application Prevention of Cell Culture Contamination
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.