missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium NCTC-109 (1X)
GREENER_CHOICE

Product Code. 10452312
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10452312

Brand: Gibco™ 21340039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for suspension and monolayer culture methods

NCTC-109 medium is a chemically defined medium that was originally developed by the National Cancer Institute and formulated for the culture of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco NCTC-109 medium is also suitable for generating and maintaining hybridoma cells. This NCTC-109 is modified as follows: With Phenol Red and Coenzymes. Without L-glutamine and HEPES. The complete formulation is available.

  • Gibco NCTC-109 medium is rich in nucleosides, vitamins, coenzymes, reducing agents and other unique ingredients
  • NCTC-109 contains no proteins, lipids, or growth factors. NCTC-109 medium typically requires supplementation of a growth factor cocktail (for serum free growth) or Fetal Bovine Serum
  • Uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hybridomas
Form Liquid
Product Type Cell Culture Medium
With Additives Phenol Red
Without Additives No Glutamine, No HEPES, No Sodium Pyruvate
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

For in vitro diagnostic use. Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.