missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, GlutaMAX™ Supplement

Product Code. 10174603
Change view
Click to view available options
Quantity:
5 L
10 L
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10174603 5 L Each
15161513 10 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174603 Supplier Gibco™ Supplier No. 61870143

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Requires

Supplementation

Originally developed to culture human leukemic cells in suspension and as a monolayer.

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas.

  • Modified as follows: with GlutaMAX™, phenol red; without HEPES
  • Contains reducing agent glutathione and high concentrations of vitamins
  • Contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium
  • Vitamins inositol and choline are present in high concentrations
  • GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (2.0g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell Type Leukemic Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives GlutaMAX, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate
Product Line Gibco, GlutaMAX, SynerGy
Quantity 5 L
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.