missing translation for 'onlineSavingsMsg'
Learn More

Gingerenone A, TRC

Product Code. 30240905
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240905 100 mg 100mg
30240805 50 mg 50mg
30240517 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240905 Supplier Toronto Research Chemicals Supplier No. TRCG387500100MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128700-97-0
Molecular Formula C21H24O5
Molecular Weight (g/mol) 356.41
Synonym (4E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-4-hepten-3-one, (E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-4-hepten-3-one;
IUPAC Name (E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hept-4-en-3-one
SMILES COc1cc(CC\C=C\C(=O)CCc2ccc(O)c(OC)c2)ccc1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H24O5/c1-25-20-13-15(8-11-18(20)23)5-3-4-6-17(22)10-7-16-9-12-19(24)21(14-16)26-2/h4,6,8-9,11-14,23-24H,3,5,7,10H2,1-2H3/b6-4+
Formula Weight 356.1624
Percent Purity >95%
Chemical Name or Material Gingerenone A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.