missing translation for 'onlineSavingsMsg'
Learn More

Girard's Reagent T, 99%

Product Code. 10616534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10616534 250 g 250g
10326750 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10616534 Supplier Thermo Scientific Acros Supplier No. 119872500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-46-6 | C5H14ClN3O | 167.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-46-6
Molecular Formula C5H14ClN3O
Molecular Weight (g/mol) 167.64
MDL Number MFCD00012009
InChI Key YSULOORXQBDPCU-UHFFFAOYSA-N
Synonym girard's reagent t, girard reagent t, girard t reagent, girards reagent t, betaine hydrazide hydrochloride, unii-oo3mf0us46, trimethylaminoacetohydrazide chloride, carbazoylmethyltrimetylammonium chloride, trimethylacethydrazide ammonium chloride, trimethylammonium chloride acethydrazide
PubChem CID 67156
SMILES [Cl-].C[N+](C)(C)CC(=O)NN

Specifications

Assay Percent Range 99%
Linear Formula H2NNHCOCH2N(CH3)3Cl
Fieser 01,410
Merck Index 15, 4465
IUPAC Name (2-hydrazinyl-2-oxoethyl)-trimethylazanium;chloride
Formula Weight 167.64
Packaging Glass bottle
Quantity 250 g
Chemical Name or Material Girard's Reagent T

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.