missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR/TNFRSF18 Antibody (CL7787) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21159PCP
This item is not returnable.
View return policy
Description
GITR/TNFRSF18 Monoclonal antibody specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot
Specifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| PerCP | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| This antibody was developed using a synthetic peptide derived from Q9Y5U5, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis | |
| 8784 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot | |
| CL7787 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction