missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gjb1 Mouse anti-Human, Rat, Clone: M12.13, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14801.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native rat Gjb1.
- High specificity
- High reactivity
Specifications
| Gjb1 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Frozen sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P08033 | |
| Gjb1 | |
| Rat junctional complexes. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| M12.13 | |
| Mouse monoclonal antibody raised against native rat Gjb1. | |
| In 10 mM PBS. | |
| Cx32 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 29584 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction