missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Glasgow MEM
BIOPHARMA

Product Code. 15323741
Change view
Click to view available options
Quantity:
50 L
Unit Size:
50 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15323741 50 L 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15323741 Supplier Corning™ Supplier No. 50020PC
  • ONLINE EXCLUSIVE PRICE
    82.35 €
    77.07 € / 50 L
    Save 5.28 €
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To assess growth promotion and cytotoxicity characteristics, Corning Classical Media products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Product performance is measured by harvest-to-plant ratios or fold increase according to predetermined specifications. Liquid and powder classical media products are also tested to determine pH and osmolality. Classical media powders are also tested for residual moisture. The pH and osmolality tests are performed according to specific procedures in which all equipment is calibrated using standards traceable to the National Institute of Standards and Technology. Biological testing of Corning liquid classical media products guarantees low levels of endotoxin, mycoplasma, and an absence of contamination. Endotoxins: classical media products are tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay. For release, each lot of standard liquid classical media must have an endotoxin level of less than 0.25 EU/mL. Classical media powder products must have an endotoxin level of less than 0.25 EU/mL when reconstituted to formula weight with low endotoxin water. All Corning liquid classical media products are tested for sterility and liquid classical media products are tested for mycoplasma off-site using a large-volume method.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes, monolayer human leukemic cells
Product Type RPMI 1640
Sterility Sterile
For Use With (Application) Mammalian Cell Culture
Quantity 50 L
Certifications/Compliance Yes
Endotoxin Level < 0.25 EU/mL
Formulation L-glutamine, inositol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.