missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLB1L3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15613925
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15613925 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613925 Supplier Invitrogen™ Supplier No. PA549990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total GLB1L3 protein.

GLB1L3 (beta-galactosidase-1-like protein 3) is a 653 amino acid protein belonging to the glycosyl hydrolase 35 family. GLB1L3 exists as three alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 11q25. With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLB1L3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GLB1L3
Gene Accession No. Q8NCI6
Gene Alias 4921509F24Rik; beta-galactosidase-1-like protein 3; galactosidase beta 1 like 3; galactosidase beta 1-like 3; galactosidase, beta 1 like 3; galactosidase, beta 1-like 3; GLB1L3
Gene Symbols GLB1L3
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human GLB1L3 (aa 291-340).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112937
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.