missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565876
This item is not returnable.
View return policy
Description
Immunogen sequence: HSPHQEPTVVN QIVPLSGERR YSMPPLFHTH YVPDIVRCVP PFREIAFLEP REITLPEAKD KLSQQILELF ETCQQQISDL KKKELCRT Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
GLD2 is cytoplasmic poly(A) RNA polymerase that adds successive AMP monomers to the 3'-end of specific RNAs, forming a poly(A) tail. In contrast to the canonical nuclear poly(A) RNA polymerase, it only adds poly(A) to selected cytoplasmic mRNAs. GLD2 does not play a role in replication-dependent histone mRNA degradation.
Specifications
| GLD2 | |
| Polyclonal | |
| Unconjugated | |
| Papd4 | |
| 8030446C20Rik; AI649009; cytoplasmic poly(A) polymerase; Gld2; GLD-2; hGLD-2; hypothetical protein LOC553626; MGC138008 protein; mGLD-2; Palp4; PAP associated domain containing 4; PAP-associated domain-containing protein 4; PAPD4; poly(A) RNA polymerase D4, non-canonical; Poly(A) RNA polymerase GLD2; Tent2; Terminal nucleotidyltransferase 2; terminal uridylyltransferase 2; TUT2; TUTase 2; TUTase2; zgc:110560 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 167153 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PIY7 | |
| Papd4 | |
| Recombinant Human GLD2. Recombinant protein control fragment (Product #RP-104999). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction