missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
219.00 € - 564.00 €
Specifications
| Antigen | GLI-1 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 25799059), In-situ Hybridization reported in scientific literature (PMID 24506883) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18237006
|
Novus Biologicals
NBP1-78259SS |
0.025 mL |
219.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18789433
|
Novus Biologicals
NBP1-78259 |
564.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
GLI-1 Polyclonal specifically detects GLI-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In-situ Hybridization.Specifications
| GLI-1 | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 25799059), In-situ Hybridization reported in scientific literature (PMID 24506883) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
| GLI1 | |
| IgG | |
| Affinity Purified |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P08151 | |
| 2735 | |
| A synthetic peptide made to an internal portion of the human Gli1 protein (between residues 150-200) [UniProt P08151] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title