Learn More
GLI-1 Antibody (CL12192) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21211AF594
Product Code. 30304868
  • 737.00 € / 0.10mL
Description
GLI-1 Monoclonal antibody specifically detects GLI-1 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GLI-1
Monoclonal
Alexa Fluor 594
GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI
This antibody was developed using a synthetic peptide derived from P08151, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
2735
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12192
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified