missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-29627SS
This item is not returnable.
View return policy
Description
GLI-3 Polyclonal specifically detects GLI-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| GLI-3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3 | |
| Rabbit | |
| 190 kDa | |
| 0.025 mg | |
| Cancer, Stem Cell Signaling Pathway | |
| 2737 | |
| Human, Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Flow Cytometry reported in scientific literature (PMID 26238488), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100, Flow (Intracellular) | |
| P10071 | |
| GLI3 | |
| A synthetic peptide made to an internal portion of human Gli3 protein (between residues 50-120) [UniProt P10071] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Immunogen has 84% homology to Xenopus and 85% homology to chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction