missing translation for 'onlineSavingsMsg'
Learn More

Glibenclamide, MedChemExpress

Product Code. 16431489
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431489 1 g 1g
16448957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431489 Supplier Medchem Express Supplier No. HY15206
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Glibenclamide (Glyburide) is an orally active ATP-sensitive K+ channel (KATP) inhibitor and can be used for the research of diabetes and obesity.

Glibenclamide inhibits P-glycoprotein. Glibenclamide directly binds and blocks the SUR1 subunits of KATP and inhibits the cystic fibrosis transmembrane conductance regulator protein (CFTR). Glibenclamide interferes with mitochondrial bioenergetics by inducing changes on membrane ion permeability. Glibenclamide can induce autophagy.

Specifications

Chemical Name or Material Glibenclamide
CAS 10238-21-8
Color White
Purity Grade Notes Research
Molecular Formula C23H28ClN3O5S
Quantity 1 g
Synonym Glyburide
Solubility Information DMSO : 250 mg/mL (506.07 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.20 mM; Need ultrasonic and warming)
SMILES ClC1=CC(C(NCCC2=CC=C(S(NC(NC3CCCCC3)=O)(=O)=O)C=C2)=O)=C(OC)C=C1
Molecular Weight (g/mol) 494.0
Formula Weight 494.0
Percent Purity 98.89%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.