missing translation for 'onlineSavingsMsg'
Learn More

Gliclazide

Product Code. 15577719 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15577719 5 g 5g
15567719 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15577719 Supplier Thermo Scientific Acros Supplier No. 461070050
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21187-98-4 | C15H21N3O3S | 323.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21187-98-4
Molecular Formula C15H21N3O3S
Molecular Weight (g/mol) 323.41
MDL Number MFCD00409893
InChI Key BOVGTQGAOIONJV-UHFFFAOYNA-N
Synonym gliclazide, glimicron, diamicron, nordialex, diaikron, gliclazida, gliklazid, glyclazide, gliclazidum inn-latin, gliclazida inn-spanish
PubChem CID 3475
ChEBI CHEBI:31654
IUPAC Name 1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-(4-methylphenyl)sulfonylurea
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=O)NN1CC2CCCC2C1

Specifications

Melting Point 163.0°C to 169.0°C
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmf, slightly soluble in methanol, ethanol
Formula Weight 323.41
Percent Purity ≥98.5% (HPLC)
Chemical Name or Material Gliclazide
compliance-icons
Product Identifier
  • Gliclazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.