missing translation for 'onlineSavingsMsg'
Learn More

Glimepiride, MedChemExpress

Product Code. 16462049
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462049 1 g 1g
16420778 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462049 Supplier Medchem Express Supplier No. HYB0104
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Glimepiride (Glimperide) is a medium-to-long acting sulfonylurea anti-diabetic compound with an ED50 of 182 μg/kg.

Glimepiride (Glimperide) is a medium-to-long acting sulfonylurea anti-diabetic compound with an ED50 of 182 μg/kg.

Specifications

Chemical Name or Material Glimepiride
CAS 93479-97-1
Color White
Purity Grade Notes Research
Molecular Formula C24H34N4O5S
Quantity 1 g
Synonym Glimperide HOE-490
Solubility Information DMSO : 33.33 mg/mL (67.93 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.20 mM; Need ultrasonic)
SMILES O=C(N1C(C(CC)=C(C)C1)=O)NCCC2=CC=C(S(=O)(NC(N[C@H]3CC[C@H](C)CC3)=O)=O)C=C2
Molecular Weight (g/mol) 490.62
Formula Weight 490.62
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.