missing translation for 'onlineSavingsMsg'
Learn More

Glipizide

Product Code. 15435649
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435649 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15435649 Supplier Thermo Scientific Alfa Aesar Supplier No. J63398.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An ATP-dependent potassium channel blocker | CAS: 29094-61-9 | C21H27N5O4S | 445.538 g/mol

An ATP-dependent potassium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29094-61-9
Color White to Yellow
Molecular Formula C21H27N5O4S
MDL Number MFCD00072159
Quantity 1 g
Merck Index 14,4442
Synonym glipizide, glucotrol, glydiazinamide, melizide, glibenese, glucozide, glupizide, sucrazide, dipazide, glupitel
Solubility Information Soluble in methanol (1.9mg/ml) and DMSO (48mg/ml)
InChI Key ZJJXGWJIGJFDTL-UHFFFAOYSA-N
SMILES CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC Name N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
Molecular Weight (g/mol) 445.538
PubChem CID 3478
ChEBI CHEBI:5384
Formula Weight 445.54
Physical Form Powder
Chemical Name or Material Glipizide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.