Learn More
Sanplatec™ Glove Box Mobile Lab Space, MBLS-2
Product Code. 30242879
Change view
Click to view available options
Material:
PVC
Unit Size:
Each
Product Code. Material Unit Size
30242879 PVC Each
1 options
  • The gloves and replacement gloves provided are simple gloves.For gas replacement, please use gas replacement gloves (Part No. 00193).
Product Code. 30242879 Supplier Sanplatec™ Supplier No. 27001
  • ONLINE EXCLUSIVE PRICE
    361.00 €
    335.28 € / Pack of 1
    Save 25.72 €
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
  • The gloves and replacement gloves provided are simple gloves.For gas replacement, please use gas replacement gloves (Part No. 00193).
Lightweight, easy-to-carry foldable glove box. Fully open front for convenient loading and unloading. Simple gas displacement is also possible.
  • The front opening is wide, making it easy to take things in and out.
  • It is easy to take on and off gloves.
  • Lightweight and foldable for easy carrying.
  • Foldable for storage.
  • It can also be used as a simple gas purge.
  • The supplied glove and replacement glove are simple disposable gloves.
  • Please use the Gas Purge glove (code 00193) for Gas Purge.
Specifications
Product Type Glove Box
Filter Type None
Lighting Type None
Material PVC
Weight (Metric) 1.7 kg