missing translation for 'onlineSavingsMsg'
Learn More

SICCO Glove Box Vitrum with Transfer Chamber

Product Code. 15735563
Change view
Click to view available options
Dimensions (L x W x H):
1210 x 700 x 600 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
15735563 1210 x 700 x 600 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15735563 Supplier SICCO Supplier No. V198608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glove Box Vitrum with Transfer Chamber Aluminium frame with panels made of borosilicate glass

  • Aluminium frame with panels made of borosilicate glass, front panel with two glove ports, right side panel and bottom side of base panel made of aluminium, top side of base panel made of stainless steel.
  • Including one pair of natural rubber gloves size 9, two lead-ins for cable on the back side (upper left and upper right).
  • Transfer chamber with two doors, external door with latch to prevent accidental opening, the tray extends into the workspace with opening of the internal door for easy removal of inserted products.
  • Work space and transfer chamber each including two hose connectors for gas-filling and aeration and appropriate tubing (2x 5 m tubing for gas-filling made of PVC, 2x 5 m aeration tubing made of PE).
  • Robust and light-weight aluminium construction
  • Superb all-round visibility
  • Non-slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
  • Connectors for gas-filling and aeration including appropriate tubing
  • Transfer chamber for inserting further products

Specifications

Product Type Glove Box
Dimensions (L x W x H) 1210 x 700 x 600 mm
Volume (Metric) 290 L
Weight (Metric) 36 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.