missing translation for 'onlineSavingsMsg'
Learn More

GLPG0492, MedChemExpress

Product Code. 16493487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493487 50 mg 50mg
16459947 5 mg 5mg
16447468 10 mg 10mg
16417436 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493487 Supplier Medchem Express Supplier No. HY18102
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GLPG0492 is a non-steroidal selective androgen receptor modulator (potency 12 nM). GLPG0492 has the potential for the research of musculo-skeletal diseases such as sarcopenia and cachexia.

GLPG0492 is a non-steroidal selective androgen receptor modulator (potency 12 nM). GLPG0492 has the potential for the research of musculo-skeletal diseases such as sarcopenia and cachexia.

Specifications

Chemical Name or Material GLPG0492
Physical Form Solid
CAS 1215085-92-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H14F3N3O3
Solubility Information DMSO : ≥ 50 mg/mL (128.43 mM)
SMILES N#CC1=CC=C(N(C(N(C)[C@]2(CO)C3=CC=CC=C3)=O)C2=O)C=C1C(F)(F)F
Molecular Weight (g/mol) 389.33
Formula Weight 389.33
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.