Learn More
Invitrogen™ Glucocorticoid Receptor Monoclonal Antibody (24GB2760)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557521
 View more versions of this product
Product Code. 30276261
  • 198.00 € / 20µL
Description
Glucocorticoid Receptor Monoclonal Antibody for Western Blot
Glucocorticoid Receptor (NR3C1) is a receptor for glucocorticoids that can act as both a transcription factor and as a regulator of other transcription factors. Glucocorticoid Receptor can also be found in heteromeric cytoplasmic complexes along with heat shock factors and immunophilins. The protein is typically found in the cytoplasm until it binds a ligand, which induces transport into the nucleus. Glucocorticoid Receptor is expressed in the heart, detected in left and right atria, left and right ventricles, aorta, apex, intraventricular septum, and atrioventricular node as well as whole adult and fetal heart. Alternate splicing, the use of at least three different promoters, and alternate translation initiation sites result in several transcript variants encoding the same protein or different isoforms, but the full-length nature of some variants has not been determined. Mutations in the Glucocorticoid Receptor gene are a cause of glucocorticoid resistance, or cortisol, resistance. Studies have shown that glucocorticoid receptor (GR) must be associated with a complex of chaperone proteins for ligand activation. GR binds to known steroids such as dexamethasone with nanomolar affinity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glucocorticoid Receptor
Monoclonal
Unconjugated
Nr3c1
cytosolic/nuclear receptor; fb13f09; GCCR; Gcr; GCRST; glucocorticoid nuclear receptor variant 1; glucocorticoid receptor; glucocorticoid receptor 1; glucocorticoid receptor alpha; GR; gr alpha; GR Kit; Grl; Grl1; Grl-1; Nr3c1; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor); OTTHUMP00000222854; OTTHUMP00000222858; utouto; utut; wu:fb13f09; zgc:113038
Mouse
Affinity Chromatography
RUO
2908
-20°C
Liquid
Western Blot
24GB2760
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P04150
Nr3c1
Recombinant protein of human Glucocorticoid Receptor (aa 1-777).
20 μL
Primary
Human
Antibody
IgG1 κ