missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucokinase/GCK Antibody (4G6), Novus Biologicals™
Mouse Monoclonal Antibody
203.00 € - 552.00 €
Specifications
| Antigen | Glucokinase/GCK |
|---|---|
| Clone | 4G6 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18184428
|
Novus Biologicals
NBP2-37579 |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653726
|
Novus Biologicals
NBP2-37579-0.025ml |
0.025 mL |
203.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glucokinase/GCK Monoclonal specifically detects Glucokinase/GCK in Human samples. It is validated for Western Blot, ELISA.Specifications
| Glucokinase/GCK | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2 | |
| GCK | |
| IgG1 | |
| Unpurified |
| 4G6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Protein Kinase | |
| Ascites with 0.03% Sodium Azide | |
| 2645 | |
| Purified recombinant fragment of human GCK expressed in E. Coli. | |
| Primary | |
| 52 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title