Learn More
Glucosylceramidase/GBA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-32271
This item is not returnable.
View return policy
Description
Glucosylceramidase/GBA Polyclonal specifically detects Glucosylceramidase/GBA in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Affinity Purified | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |