missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GluD1/GluD2 Monoclonal Antibody (848209)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561895
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Glutamate dehydrogenase (EC 1.4.1.3) catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate using NAD and/or NADP as cofactors. See also GLUD1 (MIM 138130).
Specifications
| GluD1/GluD2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P00367, P10860, P26443, P49448 | |
| GLUD1, GLUD2 | |
| E. coli-derived recombinant human GLUD1/GLUD2, Ser54-Thr558. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 848209 | |
| Unconjugated | |
| GLUD1 | |
| Ac2-281; AI118167; epididymis tissue sperm binding protein Li 18mP; GDH; GDH 1; GDH 2; GDH1; GDH2; Gdh-X; Glud; Glud1; GLUD2; Gludeha; Gludl; GLUDP1; glutamate dehydrogenase (NAD(P)+); glutamate dehydrogenase 1; glutamate dehydrogenase 1, mitochondrial; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; memory-related gene 2 protein; MRG-2; testicular secretory protein Li 14 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 14661, 24399, 2746, 2747 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction