missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR1 Antibody (S355-1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
174.00 €
Specifications
| Antigen | GluR1 |
|---|---|
| Clone | S355-1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Description
GluR1 Monoclonal specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| GluR1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252 | |
| GRIA1 | |
| IgG1 | |
| Protein G purified | |
| Detects approx. 100kDa. Does not cross-react with GluR2. |
| S355-1 | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P19490 | |
| 2890 | |
| Fusion protein amino acids 1-389 (extracellular N-terminus) of rat GluA1/GluR1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title