missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR2 Antibody (7G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37475
This item is not returnable.
View return policy
Description
GluR2 Monoclonal specifically detects GluR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| GluR2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
| Mouse | |
| 98.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 7G6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| GRIA2 | |
| Purified recombinant fragment of human GRIA2 (AA: 652-807) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 2891 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction