missing translation for 'onlineSavingsMsg'
Learn More

Glut1 Antibody (GLUT1/2475), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-75784-20ug

 View more versions of this product

Product Code. 18635378

  • 289.00 € / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Glut1 Monoclonal antibody specifically detects Glut1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glut1
Monoclonal
0.2 mg/mL
Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, ELISA 1-2 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array
P11166
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
GLUT1/2475
Unconjugated
10 mM PBS with 0.05% BSA
Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1
Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) (Uniprot: P11166)
20 μg
Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
6513
Store at 4°C.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.