missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
237.00 € - 548.00 €
Specifications
| Antigen | Glut2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1.0 - 2.0 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18726703
|
Novus Biologicals
NBP2-22218SS |
0.025 mL |
237.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18736453
|
Novus Biologicals
NBP2-22218 |
548.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Glut2 Polyclonal specifically detects Glut2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Glut2 | |
| Western Blot 1.0 - 2.0 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism, Stem Cell Signaling Pathway | |
| PBS with 0.02% Sodium Azide | |
| GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2 | |
| SLC2A2 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen sequence has 84% homology to bovine and 78% homology to rat. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P11168, P14246 | |
| 6514 | |
| A synthetic peptide made to an internal portion of the human GLUT2 protein (between residues 50-150) [UniProt# P11168] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title