missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaminyl-peptide Cyclotransferase/QPCT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00186-50ul
This item is not returnable.
View return policy
Description
Glutaminyl-peptide Cyclotransferase/QPCT Polyclonal antibody specifically detects Glutaminyl-peptide Cyclotransferase/QPCT in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Glutaminyl-peptide Cyclotransferase/QPCT | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| EC 2.3.2.5, GCT, Glutaminyl cyclase, glutaminyl-peptide cyclotransferase, Glutaminyl-tRNA cyclotransferase, Glutamyl cyclase, QCEC, sQC | |
| Produced in rabbits immunized with purified, recombinant Human Glutaminyl-peptide Cyclotransferase/QPCT (Uniprot#: Q16769-1; Ala33-Leu361) | |
| 50 μg | |
| Neuroscience | |
| 25797 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction