missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glutaredoxin 2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15974885
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15974885 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15974885 Supplier Invitrogen™ Supplier No. PA579316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A549 whole cell, rat lung tissue, mouse lung tissue. IHC: Rat Brain tissue, human lung cancer tissue, human mammary cancer tissue.

Glutaredoxin (Grx), also known as thiol transferase, is a small heat-stable oxidoreductase. Grxs form part of the glutaredoxin system, comprising NADPH, GSH and glutathione reductase, which transfers electrons from NADPH to glutaredoxins via GSH. First discovered in E. coli as GSH-dependent hydrogen donors for ribonucleotide reductase, Grx catalyzes GSH-disulfide oxidoreductase via two redox-active cysteine residues. The active sequence (Cys-Pro-Tyr-Cys) is conserved in a variety of species. The 12-kD dithiol protein has a role in reduction of mixed disulfides in cells exposed to oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutaredoxin 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.01mg sodium azide, 0.05mg thimerosal
Gene GLRX2
Gene Accession No. Q6AXW1, Q923X4, Q9NS18
Gene Alias 1700010P22Rik; AI645710; bA101E13.1 (GRX2 glutaredoxin (thioltransferase) 2); CGI-133; GLRX2; glutaredoxin (thioltransferase) 2; glutaredoxin 2; glutaredoxin 2 (thioltransferase); glutaredoxin-2, mitochondrial; GRX2
Gene Symbols GLRX2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human Glutaredoxin 2 (103-119aa EYGNQFQDALYKMTGER).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114022, 51022, 69367
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.