missing translation for 'onlineSavingsMsg'
Learn More

Glutathione S-Transferase/GST Antibody (4D12.H7.D10.E3), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-44182-0.05MG

Product Code. 18133628

  • 418.00 € / 0.05mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Glutathione S-Transferase/GST Monoclonal specifically detects Glutathione S-Transferase/GST in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoblotting.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glutathione S-Transferase/GST
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:1000, ELISA 1:50000 - 1:200000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunoblotting
Mouse
0.05 mg
Human
IgG
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoblot
4D12.H7.D10.E3
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
This Protein A purified antibody was prepared by repeated immunizations in mice with Glutathione S-Transferase/GSTProtein
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.