Learn More
Novus Biologicals™ Glutathione S-transferase Mu 5 Antibody (KAB29), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30969-100ug
 View more versions of this product
Product Code. 30067117
  • 508.00 € / 100µg
Description
Glutathione S-transferase Mu 5 Monoclonal antibody specifically detects Glutathione S-transferase Mu 5 in Broad Species Reactivity samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glutathione S-transferase Mu 5
Monoclonal
Unconjugated
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Broad Species Reactivity
Purified
Western Blot
KAB29
Western Blot : 1:1000- 1:2000
EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5
A fusion protein to the internal sequence Glutathione S-transferase Mu 5
100 μg
Signal Transduction
2949
Store at -20°C. Avoid freeze/thaw cycles.
IgG1