missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine Receptor Alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-86988
This item is not returnable.
View return policy
Description
Glycine Receptor Alpha 1 Polyclonal specifically detects Glycine Receptor Alpha 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Glycine Receptor Alpha 1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| GLRA 1, Glycine receptor 48 kDa subunit, Glycine receptor strychnine-binding subunit, glycine receptor subunit alpha-1, glycine receptor, alpha 1, glycine receptor, alpha 1 (startle disease/hyperekplexia), MGC138878, MGC138879, STHE | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GLRA1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LLRFRRKRRHHKEDEAGEGRFNFSAYGMGPACLQAKDGISVKGANNSNTTNPPPAPSKSPEEMRKLFIQRAKKIDK | |
| 0.1 mL | |
| Diabetes Research, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2741 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction