missing translation for 'onlineSavingsMsg'
Learn More

Glycitin, TRC

Product Code. 17977434
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17977434 2.5 mg 2.50mg
17938214 10 mg 10mg
17949344 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17977434 Supplier Toronto Research Chemicals Supplier No. TRCG6354102.5MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40246-10-4
Molecular Formula C22 H22 O10
Molecular Weight (g/mol) 446.4
Synonym 7-(β-D-Glucopyranosyloxy)-3-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, Glycitein 7-O-glucoside, Glycitein 7-O-β-glucoside, Glycitein-7-β-O-glucoside, Glycitin
IUPAC Name 3-(4-hydroxyphenyl)-6-methoxy-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES COc1cc2C(=O)C(=COc2cc1O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)c4ccc(O)cc4

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C22H22O10/c1-29-15-6-12-14(30-9-13(18(12)25)10-2-4-11(24)5-3-10)7-16(15)31-22-21(28)20(27)19(26)17(8-23)32-22/h2-7,9,17,19-24,26-28H,8H2,1H3/t17-,19-,20+,21-,22-/m1/s1
Formula Weight 446.1213
Percent Purity >95%
Chemical Name or Material Glycitin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.