missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycogen Phosphorylase BB/GPBB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-58760
This item is not returnable.
View return policy
Description
Glycogen Phosphorylase BB/GPBB Polyclonal antibody specifically detects Glycogen Phosphorylase BB/GPBB in Human, Mouse, Rat, Porcine, Bat, Bovine, Chicken, Hamster, Primate, Monkey, Rabbit, Sheep, Xenopus, Zebrafish samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Glycogen Phosphorylase BB/GPBB | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5834 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 10 μg/mL | |
| Brain glycogen phosphorylase, EC 2.4.1.1, Glycogen phosphorylase B, Glycogen phosphorylase brain form, Glycogen Phosphorylase Isoenzyme BB, MGC9213, Phosphorylase glycogen brain, PYGB | |
| Synthetic 20 amino acid peptide from internal region of human Glycogen Phosphorylase BB/GPBBB. | |
| 0.05 mg | |
| Primary | |
| Human, Mouse, Rat, Pig, Bat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction