missing translation for 'onlineSavingsMsg'
Learn More

Glycopyrrolate, MedChemExpress

Product Code. 16473377
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473377 50 mg 50mg
16447358 10 mg 10mg
16427326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473377 Supplier Medchem Express Supplier No. HY17465
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Glycopyrrolate (Glycopyrronium bromide) is a muscarinic competitive antagonist used as an antispasmodic.

Glycopyrrolate (Glycopyrronium bromide) is a muscarinic competitive antagonist used as an antispasmodic.

Specifications

Chemical Name or Material Glycopyrrolate
Physical Form Solid
CAS 596-51-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H28BrNO3
For Use With (Application) Neuroscience-Neuromodulation
Synonym Glycopyrronium bromideGlycopyrrolate bromide
Solubility Information H2O : 100 mg/mL (251.05 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (251.05 mM; Need ultrasonic)
SMILES C[N+]1(C)CC(OC(C(O)(C2CCCC2)C3=CC=CC=C3)=O)CC1.[Br-]
Molecular Weight (g/mol) 398.33
Formula Weight 398.33
Percent Purity 99.8%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.