missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic acid, 50% in water

Product Code. 10095720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095720 5 g Glass bottle 5g
10187840 500 g Glass bottle 500g
10300194 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10095720 Supplier Thermo Scientific Acros Supplier No. 411060050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 298-12-4 | C2H2O3 | 74.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-12-4 , 7732-18-5
Molecular Formula C2H2O3
Molecular Weight (g/mol) 74.035
MDL Number MFCD00006958
InChI Key HHLFWLYXYJOTON-UHFFFAOYSA-N
Synonym glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid
PubChem CID 760
ChEBI CHEBI:16891
IUPAC Name oxaldehydic acid
SMILES C(=O)C(=O)O

Specifications

Color Colorless to Yellow
Boiling Point 111.0°C
Melting Point -93.0°C
Linear Formula HC(O)COOH
Beilstein 03, IV, 1489
Fieser 05,320; 07,162; 09,228
Density 1.3000g/mL
Merck Index 15, 4546
Solubility Information Solubility in water: miscible.
Formula Weight 74.04
Percent Purity ≥50%
Packaging Glass bottle
Refractive Index 1.4140 to 1.4180
Specific Gravity 1.3
Quantity 5 g
Physical Form Liquid
Chemical Name or Material Glyoxylic acid, 50% in water
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.