Learn More
Glypican 3 Antibody (CL12711) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18542JF525
Product Code. 30300073
  • 770.00 € / 0.10mL
Description
Glypican 3 Monoclonal antibody specifically detects Glypican 3 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glypican 3
Monoclonal
Janelia Fluor 525
DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS
This antibody was developed using a synthetic peptide derived from P51654, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells
2719
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12711
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified